CAS RN 4091-99-0
IUPAC Name:
2-[3,6-bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic acid
Synonyms:
2-[3,6-bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic acid
Molecular Weight (g/mol):
487.29
Molecular Formula:
C24H16Cl2O7
InChi Key:
PXEZTIWVRVSYOK-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC2=C(C=C1Cl)C(C1=CC(Cl)=C(OC(C)=O)C=C1O2)C1=CC=CC=C1C(O)=O
1
–
2
of
2
results
1
2',7'-Dichlorodihydrofluorescein Diacetate, TRC
CAS: 4091-99-0 Molecular Formula: C24H16Cl2O7 Molecular Weight (g/mol): 487.29 Synonym: 2-[3,6-Bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic Acid,o-(2,7-Dichloro-3,6-dihydroxyxanthen-9-yl)benzoic Acid Diacetate,2',7'-Dichlorodihydrofluorescein Diacetate,DCFH,H2DCFDA; SMILES: CC(=O)OC1=C(C=C2C(=C1)OC3=CC(=C(C=C3C2C4=CC=CC=C4C(=O)O)Cl)OC(=O)C)Cl