accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4098-06-0

H3COOOOCH3OOCH3O

CAS RN 4098-06-0

IUPAC Name: [3,4-bis(acetyloxy)-3,4-dihydro-2H-pyran-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-3,4-dihydro-2H-pyran-2-yl]methyl acetate
Molecular Weight (g/mol): 272.25
Molecular Formula: C12H16O7
InChi Key: LLPWGHLVUPBSLP-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC=CC(OC(C)=O)C1OC(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)