accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4098-71-9

IUPAC Name: 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Synonyms: isophorone diisocyanate isophorone diamine diisocyanate IPDI
Molecular Weight (g/mol): 222.29
Molecular Formula: C12H18N2O2
InChi Key: NIMLQBUJDJZYEJ-UHFFFAOYNA-N
SMILES: CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)