accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 41014-43-1

IUPAC Name: 2-(chloromethyl)-1,3-benzoxazole
Synonyms: 2-(chloromethyl)-1,3-benzoxazole
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: ANRDUCQCZKLSGF-UHFFFAOYSA-N
SMILES: ClCC1=NC2=CC=CC=C2O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results