accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 41034-83-7

IUPAC Name: 3-(anthracen-9-yl)propanoic acid
Synonyms: 3-(anthracen-9-yl)propanoic acid
Molecular Weight (g/mol): 250.3
Molecular Formula: C17H14O2
InChi Key: BDGMYCZUEIGHJH-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results