accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 41085-43-2

IUPAC Name: 2-bromo-1-methyl-3-nitrobenzene
Synonyms: 2-bromo-1-methyl-3-nitrobenzene
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: GCAAVRIWNMTOKB-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1Br)[N+]([O-])=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results