accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 411235-57-9

IUPAC Name: cyclopropylboronic acid
Synonyms: cyclopropylboronic acid
Molecular Weight (g/mol): 85.9
Molecular Formula: C3H7BO2
InChi Key: WLVKDFJTYKELLQ-UHFFFAOYSA-N
SMILES: OB(O)C1CC1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results