accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4124-41-8

IUPAC Name: 4-methylbenzenesulfonyl 4-methylbenzene-1-sulfonate
Synonyms: 4-methylbenzenesulfonyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 326.38
Molecular Formula: C14H14O5S2
InChi Key: PDVFSPNIEOYOQL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results