CAS RN 4132-48-3
IUPAC Name:
1-methoxy-4-(propan-2-yl)benzene
Synonyms:
1-isopropyl-4-methoxybenzene
Molecular Weight (g/mol):
150.22
Molecular Formula:
C10H14O
InChi Key:
JULZQKLZSNOEEJ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C(C)C
1
–
1
of
1
results
1
4-Isopropylanisole, 95%
CAS: 4132-48-3 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.22 MDL Number: MFCD00015033 InChI Key: JULZQKLZSNOEEJ-UHFFFAOYSA-N Synonym: 4-isopropylanisole,1-isopropyl-4-methoxybenzene,4-methoxycumene,p-isopropylanisole,anisole, p-isopropyl,2-4-methoxyphenyl propane,benzene, 1-methoxy-4-1-methylethyl,1-methoxy-4-propan-2-yl benzene,p-methoxycumene,p-methoxyisopropylbenzene PubChem CID: 77783 IUPAC Name: 1-methoxy-4-propan-2-ylbenzene SMILES: COC1=CC=C(C=C1)C(C)C