accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONH2HO

CAS RN 4136-97-4

IUPAC Name: methyl 4-amino-2-hydroxybenzoate
Synonyms: methyl p-aminosalicylate pas methyl ester
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: QQOXBFUTRLDXDP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C=C1O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results