accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4141-08-6

IUPAC Name: 2-amino-N-methylbenzamide
Synonyms: 2-amino-N-methylbenzamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: KIMWOULVHFLJIU-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=CC=C1N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results