accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNBr

CAS RN 414885-80-6

IUPAC Name: 1-[(3-bromophenyl)methyl]-4-methyl-1,4-diazepane
Synonyms: 1-[(3-bromophenyl)methyl]-4-methyl-1,4-diazepane
Molecular Weight (g/mol): 283.21
Molecular Formula: C13H19BrN2
InChi Key: XUIRKBKKWOVRIJ-UHFFFAOYSA-N
SMILES: CN1CCCN(CC2=CC(Br)=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results