accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 41492-05-1

IUPAC Name: 1-bromo-4-butylbenzene
Synonyms: 1-bromo-4-butylbenzene
Molecular Weight (g/mol): 213.12
Molecular Formula: C10H13Br
InChi Key: BRGVKVZXDWGJBX-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results