CAS RN 41575-87-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
cis-Diamminedinitratoplatinum, TRC
CAS: 41575-87-5 Molecular Formula: H6N4O6Pt Molecular Weight (g/mol): 353.15 Synonym: (SP-4-2)-Diamminebis(nitrato-κO)platinum,cis-Diamminebis(nitrito-O)platinum IUPAC Name: azane;platinum(2+);dinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].N.N.[Pt+2]