accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NPtNOOOO

CAS RN 41575-94-4

IUPAC Name: carboplatin
Synonyms: carboplatin
Molecular Weight (g/mol): 373.27
Molecular Formula: C6H14N2O4Pt
InChi Key: OLESAACUTLOWQZ-UHFFFAOYSA-N
SMILES: N.N.[Pt++].OC(=O)C1(CCC1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results