accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOClNOO

CAS RN 41614-18-0

IUPAC Name: 4-chloro-N-cyclohexyl-N-methyl-3-nitrobenzamide
Synonyms: 4-chloro-N-cyclohexyl-N-methyl-3-nitrobenzamide
Molecular Weight (g/mol): 296.75
Molecular Formula: C14H17ClN2O3
InChi Key: YPYXOOOTWPRAEX-UHFFFAOYSA-N
SMILES: CN(C1CCCCC1)C(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results