accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3HNOH

CAS RN 4164-21-0

IUPAC Name: 1-phenyl-2-[(propan-2-yl)amino]ethan-1-ol
Synonyms: 2-(isopropylamino)-1-phenylethanol
Molecular Weight (g/mol): 179.26
Molecular Formula: C11H17NO
InChi Key: YANYGOBZUASRHR-UHFFFAOYNA-N
SMILES: CC(C)NCC(O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results