accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNO

CAS RN 4164-39-0

IUPAC Name: piperazine-1,4-dicarbaldehyde
Synonyms: piperazine-1,4-dicarbaldehyde
Molecular Weight (g/mol): 142.16
Molecular Formula: C6H10N2O2
InChi Key: CBLGQEBXWDKYDI-UHFFFAOYSA-N
SMILES: O=CN1CCN(CC1)C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results