accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OHH3C

CAS RN 4170-90-5

IUPAC Name: (2,4,6-trimethylphenyl)methanol
Synonyms: (2,4,6-trimethylphenyl)methanol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: LODDFDHPSIYCTK-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CO)C(C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results