accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 41716-18-1

CH3NNOO

CAS RN 41716-18-1

IUPAC Name: 1-methyl-1H-imidazole-4-carboxylate
Synonyms: 1-methylimidazole-4-carboxylate
Molecular Weight (g/mol): 125.11
Molecular Formula: C5H5N2O2
InChi Key: WZTRQGJMMHMFGH-UHFFFAOYSA-M
SMILES: CN1C=NC(=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)