accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4175-78-4

IUPAC Name: 2,5-dibromo-1,3-thiazole
Synonyms: 2,5-dibromo-1,3-thiazole
Molecular Weight (g/mol): 242.92
Molecular Formula: C3HBr2NS
InChi Key: XIBIQFJKUZZLLX-UHFFFAOYSA-N
SMILES: BrC1=CN=C(Br)S1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results