accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ON

CAS RN 4181-05-9

IUPAC Name: 4-(diphenylamino)benzaldehyde
Synonyms: 4-(diphenylamino)benzaldehyde
Molecular Weight (g/mol): 273.34
Molecular Formula: C19H15NO
InChi Key: UESSERYYFWCTBU-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results