accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4181-95-7

IUPAC Name: tetracontane
Synonyms: tetracontane
Molecular Weight (g/mol): 563.1
Molecular Formula: C40H82
InChi Key: KUPLEGDPSCCPJI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results