accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOH2NOClCl

CAS RN 418788-90-6

IUPAC Name: [3-(2,3-dichlorophenoxy)propyl](2-hydroxyethyl)azanium
Synonyms: [3-(2,3-dichlorophenoxy)propyl](2-hydroxyethyl)azanium
Molecular Weight (g/mol): 265.15
Molecular Formula: C11H16Cl2NO2
InChi Key: QVEIRCZEBQRCTR-UHFFFAOYSA-O
SMILES: OCC[NH2+]CCCOC1=CC=CC(Cl)=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results