CAS RN 418805-02-4
IUPAC Name:
ethyl 4-{4-[(5-nitrofuran-2-yl)methylidene]-3,5-dioxopyrazolidin-1-yl}benzoate
Synonyms:
ethyl 4-{4-[(5-nitrofuran-2-yl)methylidene]-3,5-dioxopyrazolidin-1-yl}benzoate
Molecular Weight (g/mol):
371.31
Molecular Formula:
C17H13N3O7
InChi Key:
ARGIPZKQJGFSGQ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=C(C=C1)N1NC(=O)C(=CC2=CC=C(O2)[N+]([O-])=O)C1=O
1
–
5
of
5
results
1
Thermo Scientific Chemicals Ubiquitin E1 Inhibitor, PYR-41
CAS: 418805-02-4 Molecular Formula: C17H13N3O7·3H2O Synonym: Ubiquitin-Activating Enzyme E1 Inhibitor; PYR-41
2
PYR-41, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
PYR-41, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
PYR-41, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.