CAS RN 41927-01-9
IUPAC Name:
3,4-dimethylbenzene-1,2-diamine
Synonyms:
3,4-dimethylbenzene-1,2-diamine
Molecular Weight (g/mol):
136.2
Molecular Formula:
C8H12N2
InChi Key:
MHQULXYNBKWNDF-UHFFFAOYSA-N
SMILES:
CC1=C(C)C(N)=C(N)C=C1
1
–
1
of
1
results
1
3,4-Dimethyl-o-phenylenediamine, 98%
CAS: 41927-01-9 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.198 MDL Number: MFCD00051709 InChI Key: MHQULXYNBKWNDF-UHFFFAOYSA-N Synonym: 3,4-dimethyl-o-phenylenediamine,2-amino-3,4-dimethylphenylamine,2-amino-3,4-dimethylphenyl amine,3,4-dimethyl-benzene-1,2-diamine,1,2-benzenediamine, 3,4-dimethyl,3,4-diamino-o-xylene,o-xylene-3,4-diamine,5,6-dimethyl phenylenediamine,1,2-diamino-3,4-dimethylbenzene,3,4-dimethyl-1,2-benzenediamine PubChem CID: 521077 IUPAC Name: 3,4-dimethylbenzene-1,2-diamine SMILES: CC1=C(C(=C(C=C1)N)N)C