CAS RN 4199-89-7
IUPAC Name:
5-chloro-1,10-phenanthroline
Synonyms:
5-chloro-1,10-phenanthroline
Molecular Weight (g/mol):
214.65
Molecular Formula:
C12H7ClN2
InChi Key:
XDUUQOQFSWSZSM-UHFFFAOYSA-N
SMILES:
ClC1=CC2=CC=CN=C2C2=NC=CC=C12
1
–
1
of
1
results
1
5-Chloro-1,10-phenanthroline
CAS: 4199-89-7 Molecular Formula: C12H7ClN2 Molecular Weight (g/mol): 214.65 MDL Number: MFCD00004980 InChI Key: XDUUQOQFSWSZSM-UHFFFAOYSA-N Synonym: 1,10-phenanthroline, 5-chloro,5-chloropyridino 3,2-h quinoline,acmc-209jmo,1,10-phenanthroline,5-chloro,5-chloro-1,10 phenanthroline,5-chloro-1,10-phenanthroline,5-chloro-1, 10-phenanthroline PubChem CID: 77865 IUPAC Name: 5-chloro-1,10-phenanthroline SMILES: ClC1=CC2=CC=CN=C2C2=NC=CC=C12