accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OCH3H3CFBFFF

CAS RN 420-37-1

IUPAC Name: boron(3+) trimethyloxidanium tetrafluoride
Synonyms: trimethyloxonium tetrafluoroborate
Molecular Weight (g/mol): 147.91
Molecular Formula: C3H9BF4O
InChi Key: KAAQTDVQNWMZJT-UHFFFAOYSA-J
SMILES: [B+3].[F-].[F-].[F-].[F-].C[O+](C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results