accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4202-14-6

IUPAC Name: dimethyl (2-oxopropyl)phosphonate
Synonyms: dimethyl 2-oxopropylphosphonate
Molecular Weight (g/mol): 166.11
Molecular Formula: C5H11O4P
InChi Key: UOWIYNWMROWVDG-UHFFFAOYSA-N
SMILES: COP(=O)(CC(C)=O)OC
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)