accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4202-38-4

H3CNCO

CAS RN 4202-38-4

IUPAC Name: 1-isocyanatododecane
Synonyms: 1-isocyanatododecane
Molecular Weight (g/mol): 211.35
Molecular Formula: C13H25NO
InChi Key: YIDSTEJLDQMWBR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results