accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFSClOO

CAS RN 421-83-0

IUPAC Name: trifluoromethanesulfonyl chloride
Synonyms: trifluoromethanesulfonyl chloride
Molecular Weight (g/mol): 168.51
Molecular Formula: CClF3O2S
InChi Key: GRGCWBWNLSTIEN-UHFFFAOYSA-N
SMILES: FC(F)(F)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)