accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4214-28-2

CH3IH3C

CAS RN 4214-28-2

IUPAC Name: 1-iodo-2,4-dimethylbenzene
Synonyms: benzene, 1-iodo-2,4-dimethyl-
Molecular Weight (g/mol): 232.06
Molecular Formula: C8H9I
InChi Key: BUNKQJAMHYKQIM-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)C(C)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)