accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 422-03-7

IUPAC Name: 2,2,3,3,3-pentafluoropropan-1-amine
Synonyms: 2,2,3,3,3-pentafluoropropan-1-amine
Molecular Weight (g/mol): 149.06
Molecular Formula: C3H4F5N
InChi Key: DPQNQLKPUVWGHE-UHFFFAOYSA-N
SMILES: NCC(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results