accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOFFFFF

CAS RN 422-05-9

IUPAC Name: 2,2,3,3,3-pentafluoropropan-1-ol
Synonyms: pentafluoro-1-propanol
Molecular Weight (g/mol): 150.05
Molecular Formula: C3H3F5O
InChi Key: PSQZJKGXDGNDFP-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results