accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOCl

CAS RN 4224-62-8

IUPAC Name: 6-chlorohexanoic acid
Synonyms: hexanoic acid, 6-chloro-
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: XWWKSLXUVZVGSP-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCl
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)