accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOBr

CAS RN 4224-70-8

IUPAC Name: 6-bromohexanoate
Synonyms: 6-bromohexanoate
Molecular Weight (g/mol): 194.05
Molecular Formula: C6H10BrO2
InChi Key: NVRVNSHHLPQGCU-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCCCCBr
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)