accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 42287-94-5

IUPAC Name: 3-(4-cyanophenyl)propanoic acid
Synonyms: 3-(4-cyanophenyl)propanoic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: RHIGOXBTMPLABF-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results