accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3OO

CAS RN 42288-46-0

IUPAC Name: 2-(3,5-dimethylphenyl)acetate
Synonyms: 2-(3,5-dimethylphenyl)acetate
Molecular Weight (g/mol): 163.2
Molecular Formula: C10H11O2
InChi Key: HDNBKTWQBJJYPD-UHFFFAOYSA-M
SMILES: CC1=CC(CC([O-])=O)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results