accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4229-44-1

IUPAC Name: hydrogen N-methylhydroxylamine chloride
Synonyms: N-methylhydroxylamine
Molecular Weight (g/mol): 83.52
Molecular Formula: CH6ClNO
InChi Key: RGZRSLKIOCHTSI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results