accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BFFFFFFK

CAS RN 42298-15-7

IUPAC Name: potassium trifluoro(trifluoromethyl)boranuide
Synonyms: potassium trifluoro(trifluoromethyl)boranuide
Molecular Weight (g/mol): 175.91
Molecular Formula: CBF6K
InChi Key: UOGBGCVSVMQVBR-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results