accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4235-95-4

IUPAC Name: (2-{[(2S)-2,3-bis[(9Z)-octadec-9-enoyloxy]propyl phosphonato]oxy}ethyl)trimethylazanium
Synonyms: (2-{[(2S)-2,3-bis[(9Z)-octadec-9-enoyloxy]propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular Weight (g/mol): 786.13
Molecular Formula: C44H84NO8P
InChi Key: SNKAWJBJQDLSFF-AOMFJDGTSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results