accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 423769-77-1

SOOSOHOCl

CAS RN 423769-77-1

IUPAC Name: 4-(4-chlorobenzenesulfonyl)thiophene-3-carboxylic acid
Synonyms: 4-(4-chlorobenzenesulfonyl)thiophene-3-carboxylic acid
Molecular Weight (g/mol): 302.74
Molecular Formula: C11H7ClO4S2
InChi Key: MSVUIONRVJKUIB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CSC=C1S(=O)(=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results