accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 42482-06-4

IUPAC Name: 3-(oct-2-en-1-yl)oxolane-2,5-dione
Synonyms: 3-(oct-2-en-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 210.27
Molecular Formula: C12H18O3
InChi Key: WSGFXVFLWVXTCJ-UHFFFAOYNA-N
SMILES: CCCCCC=CCC1CC(=O)OC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results