accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOO

CAS RN 4250-77-5

IUPAC Name: 6-hydroxy-2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: 6-hydroxyflavanone
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: XYHWPQUEOOBIOW-UHFFFAOYNA-N
SMILES: OC1=CC=C2OC(CC(=O)C2=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results