accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3

CAS RN 4250-81-1

IUPAC Name: (pent-1-yn-1-yl)benzene
Synonyms: pent-1-yn-1-ylbenzene
Molecular Weight (g/mol): 144.22
Molecular Formula: C11H12
InChi Key: DEGIOKWPYFOHGH-UHFFFAOYSA-N
SMILES: CCCC#CC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results