CAS RN 42521-82-4
IUPAC Name:
1-[(2-hydroxynaphthalen-1-yl)disulfanyl]naphthalen-2-ol
Synonyms:
1-[(2-hydroxynaphthalen-1-yl)disulfanyl]naphthalen-2-ol
Molecular Weight (g/mol):
350.45
Molecular Formula:
C20H14O2S2
InChi Key:
RFAXLXKIAKIUDT-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C=CC=CC2=C1SSC1=C(O)C=CC2=CC=CC=C12
1
–
1
of
1
results
1
Thermo Scientific Chemicals p21-Activated Kinase Inhibitor III, IPA-3
CAS: 42521-82-4 Molecular Formula: C20H14O2S2,C20H14O2S2 Synonym: PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol)