accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHOFF

CAS RN 425702-18-7

IUPAC Name: 4,5-difluoro-2-methoxybenzoic acid
Synonyms: 4,5-difluoro-2-methoxybenzoic acid
Molecular Weight (g/mol): 188.13
Molecular Formula: C8H6F2O3
InChi Key: GMFHKGUQWAQEIW-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(F)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results