accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 426-65-3

IUPAC Name: ethyl 2,2,3,3,3-pentafluoropropanoate
Synonyms: ethyl pentafluoropropionate
Molecular Weight (g/mol): 192.09
Molecular Formula: C5H5F5O2
InChi Key: DBOFMRQAMAZKQY-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results