accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClS

CAS RN 4270-70-6

IUPAC Name: triphenylsulfanium chloride
Synonyms: triphenylsulfonium chloride
Molecular Weight (g/mol): 298.83
Molecular Formula: C18H15ClS
InChi Key: ZFEAYIKULRXTAR-UHFFFAOYSA-M
SMILES: [Cl-].C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results