accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4271-13-0

CH3PHOO

CAS RN 4271-13-0

IUPAC Name: methyl(phenyl)phosphinic acid
Synonyms: methyl(phenyl)phosphinic acid
Molecular Weight (g/mol): 156.12
Molecular Formula: C7H9O2P
InChi Key: RMJCJLHZCBFPDN-UHFFFAOYNA-N
SMILES: CP(O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results